Learn More
Medchemexpress LLC D-(+)-Trehalose-d14 | 2028292-31-9 | 356.38 | 5 MG
SDP
Catalog No. 5000414565
Encompass_Preferred
D-(+)-Trehalose-d14 is the deuterium labeled D-(+)-Trehalose. D-(+)-Trehalose, isolated from *Saccharomyces cerevisiae*, can be used as a food ingredient and pharmaceutical excipient.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14565 Supplier Medchemexpress LLC Supplier No. HYN1132S5MG
D-(+)-Trehalose-d14 is the deuterium labeled D-(+)-Trehalose. D-(+)-Trehalose, isolated from *Saccharomyces cerevisiae*, can be used as a food ingredient and pharmaceutical excipient.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.