Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(+)-trehalose-d14 | 2028292-31-9 | 99.8% | 356.38 | C12H8D14O11 | 10 MG
SDP

Catalog No. 5000296574
Encompass_Preferred

D-(+)-Trehalose-d14 is the deuterium-labeled form of D-(+)-trehalose supplied as a solid analytical standard for research use. It functions as a tracer and internal standard for quantitative analyses by NMR, GC-MS, and LC-MS, and is provided with high isotopic enrichment and defined storage recommendations.

  • Deuterium-labeled trehalose with 14 deuteriums.
  • Chemical formula C12H8D14O11; molecular weight 356.38.
  • High purity: 99.8%.
  • Appearance: white to off-white solid.
  • Available in small research quantities such as 10 MG.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended use: tracer and internal standard for quantitative NMR, GC-MS, and LC-MS.

Catalog No. 50-002-96574 Supplier Medchemexpress LLC Supplier No. HYN1132S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Trehalose-d14 is the deuterium-labeled form of D-(+)-trehalose supplied as a solid analytical standard for research use. It functions as a tracer and internal standard for quantitative analyses by NMR, GC-MS, and LC-MS, and is provided with high isotopic enrichment and defined storage recommendations.

  • Deuterium-labeled trehalose with 14 deuteriums.
  • Chemical formula C12H8D14O11; molecular weight 356.38.
  • High purity: 99.8%.
  • Appearance: white to off-white solid.
  • Available in small research quantities such as 10 MG.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended use: tracer and internal standard for quantitative NMR, GC-MS, and LC-MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.