Learn More
Alkali Scientific D (+) Trehalose Dihydrate, 1 Kg
SDP
Catalog No. 5000660943
Encompass_Preferred
D (+) Trehalose Dihydrate, 1kg, is a stabilizing agent and carbohydrate compound used primarily for cell protection in laboratory applications. It is widely utilized to preserve the structural integrity of cells and proteins during freezing, dehydration, and other harsh environmental conditions. Trehalose helps to protect cellular membranes, enzymes, and nucleic acids from oxidative damage, making it essential in long-term storage of biological samples, particularly in cryopreservation and freeze-drying applications. This bulk quantity is ideal for large-scale laboratory processes or industrial applications where stable preservation of biological materials is crucial.
Catalog No. 50-006-60943 Supplier Alkali Scientific Supplier No. DTR002CP
D (+) Trehalose Dihydrate, 1kg, is a stabilizing agent and carbohydrate compound used primarily for cell protection in laboratory applications. It is widely utilized to preserve the structural integrity of cells and proteins during freezing, dehydration, and other harsh environmental conditions. Trehalose helps to protect cellular membranes, enzymes, and nucleic acids from oxidative damage, making it essential in long-term storage of biological samples, particularly in cryopreservation and freeze-drying applications. This bulk quantity is ideal for large-scale laboratory processes or industrial applications where stable preservation of biological materials is crucial.