Learn More
Sigma Aldrich Fine Chemicals Biosciences D-(+)-Trehalose dihydrate from Saccharomyces cerevisiae, >=98.5% (HPLC) | 6138-23-4 | MFCD00071594 | 100G
Catalog No. NC1224382
Encompass
D-(+)-Trehalose dihydrate from Saccharomyces cerevisiae, >=98.5% (HPLC) | Purity: >=98.5% (HPLC) | Mol Wt: 378.33 | 6138-23-4 | MFCD00071594 | 100G
Catalog No. NC1224382 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T5251100G
D-(+)-Trehalose dihydrate from Saccharomyces cerevisiae, >=98.5% (HPLC) | Purity: >=98.5% (HPLC) | Mol Wt: 378.33 | 6138-23-4 | MFCD00071594 | 100G