Learn More
Alkali Scientific D (+) Trehalose Dihydrate, 50 G
SDP
Catalog No. 5000660942
Encompass_Preferred
D (+) Trehalose Dihydrate, 50g, is a carbohydrate stabilizing agent used to protect cells, proteins, and enzymes during environmental stress such as dehydration, freezing, and oxidative conditions. Trehalose stabilizes cellular structures, preserves protein functions, and protects sensitive biological molecules from degradation. This 50g bottle is ideal for smaller-scale research projects and biochemical applications where cell protection and sample preservation are critical. It is commonly used in fields such as cryobiology, drug formulation, and the preservation of enzymes and microbial cultures during long-term storage, providing a reliable solution for stabilizing biological samples.
Catalog No. 50-006-60942 Supplier Alkali Scientific Supplier No. DTR006CP
D (+) Trehalose Dihydrate, 50g, is a carbohydrate stabilizing agent used to protect cells, proteins, and enzymes during environmental stress such as dehydration, freezing, and oxidative conditions. Trehalose stabilizes cellular structures, preserves protein functions, and protects sensitive biological molecules from degradation. This 50g bottle is ideal for smaller-scale research projects and biochemical applications where cell protection and sample preservation are critical. It is commonly used in fields such as cryobiology, drug formulation, and the preservation of enzymes and microbial cultures during long-term storage, providing a reliable solution for stabilizing biological samples.