Learn More
Medchemexpress LLC D(+)-Trehalose dihydrate | 6138-23-4 | 378.3 g/mol | 5 G
SDP
Catalog No. 5000415973
Encompass_Preferred
D-(+)-Trehalose dihydrate is an orally active disaccharide that exhibits anti-desiccant and cryopreservative activities. It functions as an osmolyte and stress protectant, aiding in carbon storage and transport. This product can be utilized as a food ingredient and pharmaceutical excipient.
  • Orally active disaccharide
  • Anti-desiccant activity
  • Cryopreservative activity
  • Acts as an osmolyte
  • Stress protectant
  • Aids in carbon storage and transport
  • Suitable for use as a food ingredient
  • Suitable for use as a pharmaceutical excipient
Catalog No. 50-004-15973 Supplier Medchemexpress LLC Supplier No. HYN1132A5G
D-(+)-Trehalose dihydrate is an orally active disaccharide that exhibits anti-desiccant and cryopreservative activities. It functions as an osmolyte and stress protectant, aiding in carbon storage and transport. This product can be utilized as a food ingredient and pharmaceutical excipient.
  • Orally active disaccharide
  • Anti-desiccant activity
  • Cryopreservative activity
  • Acts as an osmolyte
  • Stress protectant
  • Aids in carbon storage and transport
  • Suitable for use as a food ingredient
  • Suitable for use as a pharmaceutical excipient