Learn More
Medchemexpress LLC D-(+)-Trehalose dihydrate | 6138-23-4 | 378.33 | 1 G
SDP
Catalog No. 5000416614
Encompass_Preferred
D-(+)-Trehalose dihydrate is a naturally occurring disaccharide isolated from *Saccharomyces cerevisiae*. It is recognized as a safe food ingredient and pharmaceutical excipient, primarily used for research purposes.
  • Isolated from *Saccharomyces cerevisiae*
  • Can be used as a food ingredient
  • Can be used as a pharmaceutical excipient
  • For research use only
Catalog No. 50-004-16614 Supplier Medchemexpress LLC Supplier No. HYN1132A1G
D-(+)-Trehalose dihydrate is a naturally occurring disaccharide isolated from *Saccharomyces cerevisiae*. It is recognized as a safe food ingredient and pharmaceutical excipient, primarily used for research purposes.
  • Isolated from *Saccharomyces cerevisiae*
  • Can be used as a food ingredient
  • Can be used as a pharmaceutical excipient
  • For research use only