Learn More
Medchemexpress LLC D-(+)-Trehalose dihydrate | 6138-23-4 | 378.33 | 100G
SDP
Catalog No. 5000415148
Encompass_Preferred
D-(+)-Trehalose dihydrate is a safe, naturally occurring disaccharide isolated from *Saccharomyces cerevisiae*. It serves as a versatile compound with applications as both a food ingredient and a pharmaceutical excipient.
  • Isolated from Saccharomyces cerevisiae
  • Can be used as a food ingredient
  • Can be used as a pharmaceutical excipient
  • Safe and naturally occurring disaccharide
Catalog No. 50-004-15148 Supplier Medchemexpress LLC Supplier No. HYN1132A100G
D-(+)-Trehalose dihydrate is a safe, naturally occurring disaccharide isolated from *Saccharomyces cerevisiae*. It serves as a versatile compound with applications as both a food ingredient and a pharmaceutical excipient.
  • Isolated from Saccharomyces cerevisiae
  • Can be used as a food ingredient
  • Can be used as a pharmaceutical excipient
  • Safe and naturally occurring disaccharide