Learn More
Medchemexpress LLC D-(+)-trehalose dihydrate | 6138-23-4 | MFCD00071594 | 99.9% | 378.33 | C12H26O13 | 10 MG
SDP
Catalog No. 5000296576
Encompass_Preferred
D-(+)-Trehalose dihydrate analytical standard is a high-purity crystalline disaccharide reference material intended for research and analytical use. It is commonly used as a reference for qualitative and quantitative analyses and for method development in chromatography and mass spectrometry.
  • High purity suitable for analytical applications (99.9%).
  • Intended for use as a reference standard in HPLC, GC, and MS.
  • Provided as a crystalline powder isolated from Saccharomyces cerevisiae.
  • Available in small laboratory package sizes for method development and calibration.
Catalog No. 50-002-96576 Supplier Medchemexpress LLC Supplier No. HYN1132AR10MG
D-(+)-Trehalose dihydrate analytical standard is a high-purity crystalline disaccharide reference material intended for research and analytical use. It is commonly used as a reference for qualitative and quantitative analyses and for method development in chromatography and mass spectrometry.
  • High purity suitable for analytical applications (99.9%).
  • Intended for use as a reference standard in HPLC, GC, and MS.
  • Provided as a crystalline powder isolated from Saccharomyces cerevisiae.
  • Available in small laboratory package sizes for method development and calibration.