Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Trehalose dihydrate, 99%, Acros Organics™

6138-23-4
C12H26O13
378.327
DPVHGFAJLZWDOC-PVXXTIHASA-N
trehalose dihydrate,d-+-trehalose dihydrate,a,a-trehalose,d +-trehalose dihydrate,unii-7yin7j07x4,alpha,alpha-trehalose dihydrate,d-trehalose dihydrate,mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside,alp,alp.-trehalose,2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
181978
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O.O.O
6138-23-4
C12H26O13
378.327
DPVHGFAJLZWDOC-PVXXTIHASA-N
trehalose dihydrate,d-+-trehalose dihydrate,a,a-trehalose,d +-trehalose dihydrate,unii-7yin7j07x4,alpha,alpha-trehalose dihydrate,d-trehalose dihydrate,mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside,alp,alp.-trehalose,2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
181978
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O.O.O
Specific Rotation + 179.00
Melting Point 97.0°C to 99.0°C
Specific Rotation Condition + 179.00 (20.00°C c=2, H2O)
Solubility Information Solubility in water: 68.9 g/100mL (20°C). Other solubilities: soluble in hot alcohol, insoluble in ether
Formula Weight 378.32
Chemical Name or Material D(+)-Trehalose dihydrate