Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Trehalose dihydrate, >99%, MP Biomedicals™

Catalog No. ICN10309791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10309791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10309791 Supplier MP Biomedicals, Inc Supplier No. 0210309791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein stabilizing agent that functions as a cryoprotectant and protects against dehydration in biological applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
Synonym trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
PubChem CID 181978
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Color White
Assay Percent Range >99%
Quantity 1 kg
Formula Weight 378.3
Percent Purity >99%
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Trehalose dihydrate
EINECSNumber 202-739-6
Recommended Storage Store at Room Temperature (15 to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.