Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Trehalose Dihydrate, MP Biomedicals™

Catalog No. MP215394805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215394805 5 g
MP215394891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215394805 Supplier MP Biomedicals, Inc Supplier No. 0215394805
Add to Cart
Add to Cart

Used as a cryoprotectant in a variety of cell freezing media. D-(+)-Trehalose Dihydrate, MP Biomedicals is a non-reducing sugar formed from two glucose units joined by a 1-1 alpha bond. It is thought to provide plants and animals with the ability to withs

Trehalose is a natural alpha-linked, non-reducing disaccharide formed by an α,α-1,1-glucoside bond between two α-glucose units, which yields 2 moles of D-glucose upon acid hydrolysis.

  • Has 90% of the calorific content of sucrose
  • Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro
  • Oral administration of trehalose has been shown to effectively extend lifespan in a murine Huntington disease model
  • Known to accumulate in osmotically-stressed cultured bacterial cells
  • Protects against the adverse effects of dehydration
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
Synonym trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol
PubChem CID 181978
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 5 g
Solubility Information Soluble in water (50mg/mL, clear, colorless solution) or hot alcohol.
Formula Weight 378.3
Percent Purity ≥99%
Physical Form Powder
Recommended Storage Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.