Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Trehalose Dihydrate, MP Biomedicals™

Catalog No. MP210309710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210309710 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210309710 Supplier MP Biomedicals, Inc Supplier No. 0210309710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-(+)-Trehalose Dihydrate ≥98%, White Powder

  • It is a natural alpha-linked non –reducing disaccharide formed by an α,α-1,1-glucoside bond between two α-glucose units which yields 2 moles of D-glucose upon acid hydrolysis.
  • Trehalose has also been used in several biopharmaceutical monoclonal antibody formulations.
  • It is also used as a protein-stabilizing agent in research.
  • Use as a cryoprotectant in a variety of cell freezing media.
  • Trehalose does not reduce Fehling's solution; it is fermented by yeast; it is not split by alpha-glucosidase; acid hydrolysis gives 2 mols D-glucose.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
Synonym α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 203°C (literature)
Color White
Loss on Drying 8.5 - 10.5%
Quantity 10 g
Beilstein 5322018
Solubility Information Soluble in water (50mg/mL - clear, colorless solution) or hot alcohol.
Percent Purity ≥98%
Physical Form Powder
Grade ≥98%
storageNote1 Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.