Learn More
Amsbio LLC D-(+)-Trehalose Solution| 1M
Catalog No. 502041275
Encompass
Molecular Biology Grade Trehalose solution, highly characterized and stringently controlled to meet the requirements of todays nucleic acid assay systems. There is no detectable RNase or DNase activity, and endotoxin concentration meets U.S.P. requirements.
Catalog No. 50-204-1275 Supplier Amsbio LLC Supplier No. AMS.TS1M500
Molecular Biology Grade Trehalose solution, highly characterized and stringently controlled to meet the requirements of todays nucleic acid assay systems. There is no detectable RNase or DNase activity, and endotoxin concentration meets U.S.P. requirements.