Learn More
Medchemexpress LLC [D-Trp34]-Neuropeptide Y | 153549-84-9 | 99.7% | 4311.72 | 5 MG
SDP
Catalog No. 5000424567
Encompass_Preferred
[D-Trp34]-Neuropeptide Y is a potent and selective neuropeptide Y (NPY) Y5 receptor agonist, showing significantly less potency at NPY Y1, Y2, Y4, and y6 receptors. This compound is known to markedly increase food intake in rats.
  • Potent and selective NPY Y5 receptor agonist
  • Significantly less potent at NPY Y1, Y2, Y4, and y6 receptors
  • Increases food intake in rats
  • Soluble in water at 100 mg/mL (23.19 mM)
  • Appears as a white to off-white solid
Catalog No. 50-004-24567 Supplier Medchemexpress LLC Supplier No. HYP13225MG
[D-Trp34]-Neuropeptide Y is a potent and selective neuropeptide Y (NPY) Y5 receptor agonist, showing significantly less potency at NPY Y1, Y2, Y4, and y6 receptors. This compound is known to markedly increase food intake in rats.
  • Potent and selective NPY Y5 receptor agonist
  • Significantly less potent at NPY Y1, Y2, Y4, and y6 receptors
  • Increases food intake in rats
  • Soluble in water at 100 mg/mL (23.19 mM)
  • Appears as a white to off-white solid