Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (D-Trp6)-LHRH free acid | 129418-54-8 | 99.67% | 1312.43 | 1 MG
SDP

Catalog No. 5000396144
Encompass_Preferred

(D-Trp6)-LHRH free acid is a luteinizing hormone-releasing hormone (LHRH) agonist. It has been shown in vitro to reduce the proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27.

  • Luteinizing hormone-releasing hormone (LHRH) agonist
  • Reduces proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27
  • Purity of 99.67%
  • Molecular weight of 1312.43
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-96144 Supplier Medchemexpress LLC Supplier No. HYP45721MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(D-Trp6)-LHRH free acid is a luteinizing hormone-releasing hormone (LHRH) agonist. It has been shown in vitro to reduce the proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27.

  • Luteinizing hormone-releasing hormone (LHRH) agonist
  • Reduces proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27
  • Purity of 99.67%
  • Molecular weight of 1312.43
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.