Learn More
Medchemexpress LLC (D-Trp6)-LHRH free acid | 129418-54-8 | 99.7% | 1312.43 | 10 MG
SDP
Catalog No. 5000398553
Encompass_Preferred
(D-Trp6)-LHRH free acid is a luteinizing hormone-releasing hormone (LHRH) agonist. In vitro, (D-Trp6)-LHRH free acid (10 μM; 24 h or 6 days) reduces the proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27.
  • Acts as a luteinizing hormone-releasing hormone (LHRH) agonist
  • Reduces proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27
  • High purity of 99.67%
  • White to off-white solid appearance
Catalog No. 50-003-98553 Supplier Medchemexpress LLC Supplier No. HYP457210MG
(D-Trp6)-LHRH free acid is a luteinizing hormone-releasing hormone (LHRH) agonist. In vitro, (D-Trp6)-LHRH free acid (10 μM; 24 h or 6 days) reduces the proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27.
  • Acts as a luteinizing hormone-releasing hormone (LHRH) agonist
  • Reduces proliferation of human epithelial ovarian cancer cell lines EFO-21 and EFO-27
  • High purity of 99.67%
  • White to off-white solid appearance