Learn More
Medchemexpress LLC [D-Trp8]-γ-MSH | 321351-81-9 | 98.3% | 1570.77 | 1 MG
SDP
Catalog No. 5000424500
Encompass_Preferred
[D-Trp8]-γ-MSH is a potent and selective agonist of the melanocortin 3 (MC3) receptor. It exhibits IC50 values of 6.7 nM for hMC3, 600 nM for hMC4, and 340 nM for hMC5 in CHO cells. This compound may offer protection against various inflammatory disorders, such as rheumatoid arthritis and colitis.
  • Potent and selective agonist of melanocortin 3 (MC3) receptor
  • Offers protection against multiple inflammatory disorders
  • IC50 of 6.7 nM for hMC3
  • IC50 of 600 nM for hMC4
  • IC50 of 340 nM for hMC5
  • Solid, white to off-white appearance
  • Soluble in H2O at 12.5 mg/mL (requires ultrasonic)
Catalog No. 50-004-24500 Supplier Medchemexpress LLC Supplier No. HYP12171MG
[D-Trp8]-γ-MSH is a potent and selective agonist of the melanocortin 3 (MC3) receptor. It exhibits IC50 values of 6.7 nM for hMC3, 600 nM for hMC4, and 340 nM for hMC5 in CHO cells. This compound may offer protection against various inflammatory disorders, such as rheumatoid arthritis and colitis.
  • Potent and selective agonist of melanocortin 3 (MC3) receptor
  • Offers protection against multiple inflammatory disorders
  • IC50 of 6.7 nM for hMC3
  • IC50 of 600 nM for hMC4
  • IC50 of 340 nM for hMC5
  • Solid, white to off-white appearance
  • Soluble in H2O at 12.5 mg/mL (requires ultrasonic)