Learn More
Cayman Chemical [D-Trp8]-γ-MSH (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502953542
Encompass_Preferred
An MC3R agonist; selectively induces cAMP accumulation in cells expressing MC3R over cells expressing MC4R or MC5R (EC50s = 0.33, 100, and 82 nM, respectively, for the human receptors); reduces the incidence of ventricular tachycardia, ventricular fibrillation, and death in a rat model of myocardial ischemia-reperfusion injury at 162 nmol/kg, i.v.; inhibits urate-induced CXCL1 release and peritoneal polymorphonuclear leukocyte accumulation in recessive yellow (e/e) mice with a non-functional MC1R
Catalog No. 50-295-3542 Supplier Cayman Chemical Supplier No. 1403710 MG
An MC3R agonist; selectively induces cAMP accumulation in cells expressing MC3R over cells expressing MC4R or MC5R (EC50s = 0.33, 100, and 82 nM, respectively, for the human receptors); reduces the incidence of ventricular tachycardia, ventricular fibrillation, and death in a rat model of myocardial ischemia-reperfusion injury at 162 nmol/kg, i.v.; inhibits urate-induced CXCL1 release and peritoneal polymorphonuclear leukocyte accumulation in recessive yellow (e/e) mice with a non-functional MC1R