Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Tryptophan Benzyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T24381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24381G Supplier TCI America Supplier No. T24381G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 22839-16-3
Molecular Formula C18H19ClN2O2
Molecular Weight (g/mol) 330.812
MDL Number MFCD00237293
InChI Key DOKDMGOWZOTZRA-PKLMIRHRSA-N
Synonym H-D-Trp-OBzl.HCl
PubChem CID 44629900
IUPAC Name benzyl (2R)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)C(CC2=CNC3=CC=CC=C32)N.Cl

Specifications

Melting Point 215°C
Color White-Yellow
Quantity 1 g
Formula Weight 330.81
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material D-Tryptophan Benzyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.