Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ D-Tube™ Dialyzer Midi, MWCO 3.5 kDa

Catalog No. 715063 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10d-tu
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-506-3 10d-tu
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-506-3 Supplier MilliporeSigma™ Supplier No. 715063
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for dialysis and electroelution of proteins, RNA, DNA, and oligonucleotides from polyacrylamide or agarose gels

  • Easy-to-handle dialyzers for buffer exchange and removal of urea, detergents
  • Ideal for electroelution of proteins, protein-DNA complexes, oligonucleotides, DNA, and RNA from polyacrylamide and agarose gels
  • One-step procedure that does not require syringes or any special equipment
  • Sample volume recovery > 97%
  • Protease, RNAse, DNAse, and PCR product free
TRUSTED_SUSTAINABILITY

Specifications

Product Type D-Tube Dialyzer Midi, MWCO 3.5 kDa
Quantity 10d-tu
Includes
  • 10 D-Tube Dialyzers and one floating rack that can hold up to four D-Tube Dialyzers
Content And Storage +15°C to +30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.