Learn More
MilliporeSigma™ D-Tube™ Electroelution Accessory Kit
Catalog No. 715113 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
71-511-3 1 kit
1 options
Catalog No. 71-511-3 Supplier MilliporeSigma™ Supplier No. 715113
Add to Cart
Add to Cart
Life Science research application
D-Tube Electroelution Accessory Kit provides supporting trays fitting all three D-Tube Dialyzers (Mini, Midi, and Maxi) and that are compatible with most commercially-available horizontal electrophoresis units. Reagents are also provided for protein and nucleic acid precipitation following electroelution. In addition, the kit contains buffer optimized for thorough removal of SDS from the extracted protein samples, rendering them compatible with MALDI-MS analysis.
TRUSTED_SUSTAINABILITY
Specifications
Product Type D-Tube Electroelution Accessory Kit
Quantity 1 kit
Content And Storage +15°C to +30°C
Column Type Column Accesory