Learn More
MilliporeSigma™ D-Tube96™ Dialyzer, 12-14 kDa
Catalog No. 717133 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
96d-tu
Catalog No. Quantity
71-713-3 96d-tu
1 options
Catalog No. 71-713-3 Supplier MilliporeSigma™ Supplier No. 717133
Add to Cart
Add to Cart
Convenient and simultaneous dialysis of 96 samples in high throughput manner
  • The device takes advantage of the features of the D-Tube™ Dialyzer Mini in a 96-tube configuration.
  • Easy to handle dialyzers with dual membranes and large surface area
  • The membrane is ultra clean, EDTA-treated regenerated cellulose, sulfur- and heavy metal- free and available in molecular weight cutoffs of 6-8 kDa or 12-14 kDa
  • The D-Tube Dialyzers Mini have a capacity of 10-250μL and are ideal for small samples
  • The samples are added and removed using a standard laboratory pipet, multichannel pipet, or by robotic dispensing
  • Suitable for dialysis of proteins, oligonucleotide, RNA, and DNA in a high-throughput format
  • Each kit contains a floating rack with 96 D-Tube Dialyzers Mini, 96 caps and an adhesive aluminum plate sealer
  • If fewer than 96 tubes are required, excess tubes can be removed from the modular device and saved for use at a later time
  • Ideally suited for buffer exchange of protein samples into a physiologically relevant buffer after screening for optimal refolding conditions with the iFOLD™ Protein Refolding Systems
TRUSTED_SUSTAINABILITY
Specifications
Product Type D-Tube96 Dialyzer, 12-14 kDa
Quantity 96d-tu
Includes A floating rack with 96 D-Tube Dialyzers Mini, 96 caps and an adhesive aluminum plate sealer
Content And Storage +15°C to +30°C