Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich D-Tyrosine

Catalog No. 855456500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-545-6500MG 5 g
85-545-65G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 85-545-6500MG Supplier Sigma Aldrich Supplier No. 855456500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

D-Tyrosine

Specifications

CAS 556-02-5
Melting Point >300°C
Molecular Formula C9H11NO3
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
Quantity 5 g
MDL Number MFCD00063073
Synonym (R)-2-Amino-3-(4-hydroxyphenyl)propionic acid; 3-(4-Hydroxyphenyl)-D-alanine
Molecular Weight (g/mol) 181.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.