Learn More
Medchemexpress LLC HY-Y0444 500mg Medchemexpress, D-Tyrosine CAS:556-02-5 Purity:>98%
Supplier: Medchemexpress LLC HYY0444500MG
Medchemexpress, HY-Y0444 500mg D-Tyrosine CAS:556-02-5 D-Tyrosine is the D-isomer of tyrosine. D-Tyrosine negatively regulates melanin synthesis by inhibiting tyrosinase activity. D-Tyrosine inhibits biofilm formation and trigger the self-dispersal of biofilms without suppressing bacterial growth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.