Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Tyrosine, 98%

Catalog No. AC226910100
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226910100 10 g Glass Bottle
AC226910025 2.5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226910100 Supplier Thermo Scientific Chemicals Supplier No. 226910100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 556-02-5 | C9H11NO3 | 181.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-02-5
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063073
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym d-tyrosine, h-d-tyr-oh, d-tyr, r-2-amino-3-4-hydroxyphenyl propanoic acid, d-tyrosin, 2r-2-amino-3-4-hydroxyphenyl propanoic acid, 3-4-hydroxyphenyl-d-alanine, d-p-tyrosine, tyrosine, d, d-+-tyrosine
PubChem CID 71098
ChEBI CHEBI:28479
IUPAC Name (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C, 3 hrs)
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
Residue after Ignition 0.1% max.
Beilstein 14, 605
Specific Rotation +10° to +12° (20°C, 589nm) (c=5, 1N HCl) (after 2 hrs.)
Merck Index 14, 9839
Specific Rotation Condition +11.30° (20°C c=5,1N HCl)
Quantity 10 g
Formula Weight 181.19
Percent Purity ≥97.5%
Physical Form Powder or Crystals
Chemical Name or Material D-Tyrosine
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.