Learn More
Medchemexpress LLC D-tyrosine-d4 | 944386-47-4 | 99.0% | 185.21 g/mol | C9H7D4NO3 | 5mg
SDP
Catalog No. 5000270201
Encompass_Preferred
D-Tyrosine-d4 is the deuterium labeled D-Tyrosine (HY-Y0444)[1] D-Tyrosine is the D-isomer of tyrosine D-Tyrosine negatively regulates melanin synthesis by inhibiting tyrosinase activity D-Tyrosine inhibits biofilm formation and trigger the self-dispersal of biofilms without suppressing bacterial growth[2][3]
Catalog No. 50-002-70201 Supplier Medchemexpress LLC Supplier No. HYY0444S15MG
D-Tyrosine-d4 is the deuterium labeled D-Tyrosine (HY-Y0444)[1] D-Tyrosine is the D-isomer of tyrosine D-Tyrosine negatively regulates melanin synthesis by inhibiting tyrosinase activity D-Tyrosine inhibits biofilm formation and trigger the self-dispersal of biofilms without suppressing bacterial growth[2][3]