Learn More
Medchemexpress LLC D-tyrosine-d7 | 1426174-46-0 | 97.3% | 188.23 | C9H4D7NO3 | 10mg
SDP
Supplier:  Medchemexpress LLC HYY0444S210MG
Encompass_Preferred
D-Tyrosine-d7 is the deuterium labeled D-Tyrosine (HY-Y0444)[1] D-Tyrosine is the D-isomer of tyrosine D-Tyrosine negatively regulates melanin synthesis by inhibiting tyrosinase activity D-Tyrosine inhibits biofilm formation and trigger the self-dispersal of biofilms without suppressing bacterial growth[2][3]
Catalog No. 50-002-69266