Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Guardair Static Conductive Drum Vacuum, 30 Gallon, Air-Powered, Steel

Catalog No. 502838968
Encompass

This air-powered, static conductive drum vacuum effectively vacuums chips, dust, debris, liquids, or sludges. Its electrically bonded components and conductive attachments prevent static discharge, ensuring safe and efficient performance with exceptional vacuum lift and flow.

  • Single Venturi vacuum head with exhaust silencer
  • Heavy-duty, static conductive, powder-coated 30 gallon steel drum
  • Standard cartridge filter
  • Air shutoff valve (3/4 in FNPT inlet, universal couplings)
  • Detachable drum dolly (swivel casters, 3 in steel wheels)
  • Static conductive air hose (3/4 in x 20 ft, universal couplings)
  • Static conductive vacuum hose (20 ft)
  • Detachable 12 in wide metal wet/dry floor tool with 4 ft steel wand
  • 24 in metal crevice tool
  • Factory-installed options: Liquid Shutoff, HEPA Filtration, Extra Silent Exhaust

Catalog No. 50-283-8968 Supplier MSC Supplier No. 39466826
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This air-powered, static conductive drum vacuum effectively vacuums chips, dust, debris, liquids, or sludges. Its electrically bonded components and conductive attachments prevent static discharge, ensuring safe and efficient performance with exceptional vacuum lift and flow.

  • Single Venturi vacuum head with exhaust silencer
  • Heavy-duty, static conductive, powder-coated 30 gallon steel drum
  • Standard cartridge filter
  • Air shutoff valve (3/4 in FNPT inlet, universal couplings)
  • Detachable drum dolly (swivel casters, 3 in steel wheels)
  • Static conductive air hose (3/4 in x 20 ft, universal couplings)
  • Static conductive vacuum hose (20 ft)
  • Detachable 12 in wide metal wet/dry floor tool with 4 ft steel wand
  • 24 in metal crevice tool
  • Factory-installed options: Liquid Shutoff, HEPA Filtration, Extra Silent Exhaust

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.