Learn More
MSC Guardair Vacuum Kit, 10 gal., D Venturi, 1 ea
Catalog No. 502869713
Encompass
The Guardair 10 gal. COMP VAC™ Kits offer high performance in a compact, lightweight, and portable package. These kits are designed for vacuuming up chips, dust, debris, liquids, or sludge, powered by compressed air for dependable and trouble-free operation.
  • Single venturi vacuum generating head with exhaust silencer
  • Standard cartridge filter
  • Canister style steel drum with carry handles
  • Air shutoff valve with 1/2 in FNPT inlet
  • Standard-duty vinyl vacuum hose, 1.5 in x 20 ft
  • Detachable 12 in wide metal wet/dry floor tool
Catalog No. 50-286-9713 Supplier MSC Supplier No. 49241516
The Guardair 10 gal. COMP VAC™ Kits offer high performance in a compact, lightweight, and portable package. These kits are designed for vacuuming up chips, dust, debris, liquids, or sludge, powered by compressed air for dependable and trouble-free operation.
  • Single venturi vacuum generating head with exhaust silencer
  • Standard cartridge filter
  • Canister style steel drum with carry handles
  • Air shutoff valve with 1/2 in FNPT inlet
  • Standard-duty vinyl vacuum hose, 1.5 in x 20 ft
  • Detachable 12 in wide metal wet/dry floor tool