Learn More
MSC GuardAir Pneumatic Drum Vacuum Kit, ATEX, 55 gal, 1 ea
Catalog No. 502817470
Encompass
This pneumatic drum vacuum kit is designed for vacuuming chips, dust, debris, liquids, or sludge in hazardous locations. Powered by compressed air, it operates without motors or electric cords, preventing sparking or burnout. It generates exceptional vacuum lift and flow, providing safe, effective, and efficient performance.
  • Electrically bonded components and static conductive attachments
  • Multiple grounding options eliminate static electricity at the source
  • Heavy-duty, static conductive powder coated, 55 gal steel drum
  • Single venturi vacuum generating head with exhaust silencer
  • Standard cartridge filter with electrically conductive cage
  • Detachable drum dolly with four, 3 in steel swivel casters
  • ATEX certified
Catalog No. 50-281-7470 Supplier MSC Supplier No. 92383421
This pneumatic drum vacuum kit is designed for vacuuming chips, dust, debris, liquids, or sludge in hazardous locations. Powered by compressed air, it operates without motors or electric cords, preventing sparking or burnout. It generates exceptional vacuum lift and flow, providing safe, effective, and efficient performance.
  • Electrically bonded components and static conductive attachments
  • Multiple grounding options eliminate static electricity at the source
  • Heavy-duty, static conductive powder coated, 55 gal steel drum
  • Single venturi vacuum generating head with exhaust silencer
  • Standard cartridge filter with electrically conductive cage
  • Detachable drum dolly with four, 3 in steel swivel casters
  • ATEX certified