Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ D-Val-Leu-Lys-7-amino-4-methylcoumarin

Catalog No. MP3AMC08110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP3AMC08110 10mg
MP3AMC08125 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP3AMC08110 Supplier MP Biomedicals™ Supplier No. 03AMC08110
Add to Cart
Add to Cart

D-Val-Leu-Lys-7-amino-4-methylcoumarin

  • It is used for assay of tissue plasminogen activator.
  • The AMC fluorogenic substrates are widely used for routine assays of peptidases.
  • A short peptide sequence is conjugated to 7-AMINO-4-METHYLCOUMARIN, which is released upon cleavage by peptidase.
  • The release of 7-AMINO-4-METHYLCOUMARIN is then measured spectrophotometrically.
  • The D-VAL-LEU-LYS-7-AMINO-4-METHYLCOUMARIN is used as s substrate for Plasmin
TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Solid
For Use With (Application) Apoptosis, Cell Viability Analysis, Cell Signalling
Quantity 10mg
Molecular Weight (g/mol) 515.4 (Not including 2TFA salt)
Product Type Proteins, Enzymes & Peptides
Storage Requirements Store at Room Temperature (15-30°C). Keep cool and dry.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.