Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Val-Phe-Lys-CMK (diTFA) | 97.4% | 653.01 | 10 MG
SDP

Catalog No. 5000424624
Encompass_Preferred

D-Val-Phe-Lys-CMK (diTFA) is a chloromethyl ketone derivative that functions as an inhibitor of plasmin activity. This chemical compound is primarily intended for research use in laboratory settings for the manufacture of substances and has not been fully validated for medical applications.

  • Inhibits plasmin activity
  • Molecular formula: C25H35ClF6N4O7
  • Appearance: white to off-white solid
  • Targets ser/thr protease
  • Stable under recommended storage conditions

Catalog No. 50-004-24624 Supplier Medchemexpress LLC Supplier No. HYP10095A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Val-Phe-Lys-CMK (diTFA) is a chloromethyl ketone derivative that functions as an inhibitor of plasmin activity. This chemical compound is primarily intended for research use in laboratory settings for the manufacture of substances and has not been fully validated for medical applications.

  • Inhibits plasmin activity
  • Molecular formula: C25H35ClF6N4O7
  • Appearance: white to off-white solid
  • Targets ser/thr protease
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.