Learn More
Medchemexpress LLC D-Valinol | 4276-09-9 | 99.7% | 103.16 | 25 G
SDP
Catalog No. 5000402955
Encompass_Preferred
(2R)-2-Amino-3-methylbutan-1-ol is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used as a biochemical reagent
  • Functions as a biological material or organic compound
  • Suitable for life science research
  • Soluble in DMSO at 100 mg/mL
  • Pure form is stable for 3 years at -20°C
  • In solvent, it is stable for 6 months at -80°C
Catalog No. 50-004-02955 Supplier Medchemexpress LLC Supplier No. 25GHY30167
(2R)-2-Amino-3-methylbutan-1-ol is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used as a biochemical reagent
  • Functions as a biological material or organic compound
  • Suitable for life science research
  • Soluble in DMSO at 100 mg/mL
  • Pure form is stable for 3 years at -20°C
  • In solvent, it is stable for 6 months at -80°C