Learn More
Essendant D-Vour Absorbent Powder, Canister, Lemon, 16oz, 6/Carton
Catalog No. 502002575
Encompass_Preferred
BIG D D-VOUR devours liquids such as spilled bodily fluids as it eliminates odor. D-VOUR is a polymer which, when sprinkled lightly on liquid, transforms it in seconds to a tidy, congealed semi-solid, easy to pick up and dispose of. D-VOUR will absorb 100 times its weight. D-VOUR changes smelly liquids into an odorless semi-solid mass for fast, easy clean-up. Ideal for spills in grocery stores, restaurants, health care facilities or schools. Use D-VOUR to set free-flowing liquids (except for oils) and destroy odors fast.
Catalog No. 50-200-2575 Supplier Essendant Supplier No. BGD166
BIG D D-VOUR devours liquids such as spilled bodily fluids as it eliminates odor. D-VOUR is a polymer which, when sprinkled lightly on liquid, transforms it in seconds to a tidy, congealed semi-solid, easy to pick up and dispose of. D-VOUR will absorb 100 times its weight. D-VOUR changes smelly liquids into an odorless semi-solid mass for fast, easy clean-up. Ideal for spills in grocery stores, restaurants, health care facilities or schools. Use D-VOUR to set free-flowing liquids (except for oils) and destroy odors fast.