Learn More
Bellco Glass, Inc. The Bellco Refrigerated Roll-In Incubators have 40 cubic feet of volume and are designed for use with Bellco's Cell Production Roller Apparatus (CPRA). The flush-to-floor design allows easy movement of the CPRA in and out of the chamber. Four internal c
SDP
Catalog No. 501946055
Encompass_Preferred
The Bellco Refrigerated Roll-In Incubators have 40 cubic feet of volume and are designed for use with Bellco's Cell Production Roller Apparatus (CPRA). The flush-to-floor design allows easy movement of the CPRA in and out of the chamber. Four internal c
Catalog No. 50-194-6055 Supplier Bellco Glass, Inc. Supplier No. 772851005
Add to Cart
Add to Cart
Request a Quote
The Bellco Refrigerated Roll-In Incubators have 40 cubic feet of volume and are designed for use with Bellco's Cell Production Roller Apparatus (CPRA). The flush-to-floor design allows easy movement of the CPRA in and out of the chamber. Four internal c