Learn More
Bellco Glass, Inc. Double Window Roll-In Incubator, 230V
SDP
Catalog No. 501946059
Encompass_Preferred
Bellco Roll-In Incubators have 40 cubic feet of volume and are designed for use with Bellco’s Cell Production Roller Apparatus (CPRA). The flush-to-floor design allows easy movement of the CPRA in and out of the chamber. Four internal convenience outlets allow simple connection to internally placed appliances such as CPRA, Roller Drums, Magnetic Stirrers, etc. Fans take air from the chamber top, pass it over the 1100 watt heater and re-inject it into the chamber on the bottom back wall. A microprocessor temperature controller and membrane keypad allows simple setting and monitoring of chamber temperature. A safety thermostat is included to prevent over-temperature failures. Shelf brackets are included for mounting of optional shelves. The control panel features indicating lights and dials for automatic control of temperature.. MADE IN THE USA
Catalog No. 50-194-6059 Supplier Bellco Glass, Inc. Supplier No. 772859522
Add to Cart
Add to Cart
Request a Quote
Bellco Roll-In Incubators have 40 cubic feet of volume and are designed for use with Bellco’s Cell Production Roller Apparatus (CPRA). The flush-to-floor design allows easy movement of the CPRA in and out of the chamber. Four internal convenience outlets allow simple connection to internally placed appliances such as CPRA, Roller Drums, Magnetic Stirrers, etc. Fans take air from the chamber top, pass it over the 1100 watt heater and re-inject it into the chamber on the bottom back wall. A microprocessor temperature controller and membrane keypad allows simple setting and monitoring of chamber temperature. A safety thermostat is included to prevent over-temperature failures. Shelf brackets are included for mounting of optional shelves. The control panel features indicating lights and dials for automatic control of temperature.. MADE IN THE USA