Learn More
Cayman Chemical D-(+)-Xylose, Purity: ≥95%, CAS: 58-86-6, 100 g
SDP
Catalog No. 502947489
Encompass_Preferred
A monosaccharide; has commonly been used as a starting material in the commercial biosynthesis of xylitol and biofuels
Catalog No. 50-294-7489 Supplier Cayman Chemical Supplier No. 33236100 G
A monosaccharide; has commonly been used as a starting material in the commercial biosynthesis of xylitol and biofuels