Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-(+)-xylose-2-13C | 131771-69-2 | 99.9% | 151.12 | C5H10O5 (13C at C-2) | 10 MG
SDP

Catalog No. 5000298046
Encompass_Preferred

Xylose-2-13C is a 13C-labelled form of D-xylose provided as a solid tracer and analytical standard for metabolic and analytical studies. It is suitable as an internal standard or tracer for NMR, GC-MS, and LC-MS quantitative analyses and can be used in biochemical or microbial fermentation research as a carbon source.

  • Contains 10 mg of material.
  • Purity 99.89%.
  • Isotope labeling: 13C at the 2-position.
  • CAS number 131771-69-2.
  • Molecular weight 151.12 g/mol.
  • Solubility: in DMSO ~50 mg/mL; requires ultrasonic and warming; hygroscopic DMSO affects solubility.
  • Storage: store protected from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Appearance: white to off-white solid.
  • Intended uses include tracer studies and use as an internal standard in analytical workflows.

Catalog No. 50-002-98046 Supplier Medchemexpress LLC Supplier No. HYN0537S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xylose-2-13C is a 13C-labelled form of D-xylose provided as a solid tracer and analytical standard for metabolic and analytical studies. It is suitable as an internal standard or tracer for NMR, GC-MS, and LC-MS quantitative analyses and can be used in biochemical or microbial fermentation research as a carbon source.

  • Contains 10 mg of material.
  • Purity 99.89%.
  • Isotope labeling: 13C at the 2-position.
  • CAS number 131771-69-2.
  • Molecular weight 151.12 g/mol.
  • Solubility: in DMSO ~50 mg/mL; requires ultrasonic and warming; hygroscopic DMSO affects solubility.
  • Storage: store protected from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Appearance: white to off-white solid.
  • Intended uses include tracer studies and use as an internal standard in analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.