Learn More
Cayman Chemical D-(+)-Xylose, Purity: ≥95%, CAS: 58-86-6, 250 g
SDP
Catalog No. 502948069
Encompass_Preferred
A monosaccharide; has commonly been used as a starting material in the commercial biosynthesis of xylitol and biofuels
Catalog No. 50-294-8069 Supplier Cayman Chemical Supplier No. 33236250 G
A monosaccharide; has commonly been used as a starting material in the commercial biosynthesis of xylitol and biofuels