Learn More
Sigma Aldrich Fine Chemicals Biosciences D-(+)-Xylose, 58-86-6, MFCD00064360, 500 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences X1500500G
Encompass
Empirical Formula (Hill Notation): C5H10O5, Molecular Weight: 150.13, ≥99%, mp: 154-158 °C.
Catalog No. NC0750134