Learn More
Medchemexpress LLC α-D-Xylose | 6763-34-4 | 150.13 | 1 KG
SDP
Catalog No. 5000415995
Encompass_Preferred
α-D-Xylose is a basic component of the five-carbon fraction of biomass and a precursor of hemicellulose. It participates in various enzyme-catalyzed reactions and metabolic pathways. It is also used in tanning, dyeing, and as a diabetic food. The product appears as a white to off-white solid and is for research use only.
  • Basic component of the five-carbon fraction of biomass
  • Precursor of hemicellulose
  • Participates in various enzyme-catalyzed reactions and metabolic pathways
  • Used in tanning, dyeing, and as a diabetic food
  • White to off-white solid appearance
  • Intended for research use only
Catalog No. 50-004-15995 Supplier Medchemexpress LLC Supplier No. HYN20181KG
α-D-Xylose is a basic component of the five-carbon fraction of biomass and a precursor of hemicellulose. It participates in various enzyme-catalyzed reactions and metabolic pathways. It is also used in tanning, dyeing, and as a diabetic food. The product appears as a white to off-white solid and is for research use only.
  • Basic component of the five-carbon fraction of biomass
  • Precursor of hemicellulose
  • Participates in various enzyme-catalyzed reactions and metabolic pathways
  • Used in tanning, dyeing, and as a diabetic food
  • White to off-white solid appearance
  • Intended for research use only