Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ D (+) Xylose
SCI_ED

Catalog No. s25630 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25630 25 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25630
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 58-86-6
Synonym Wood Sugar

Specifications

Density 1.525 at 20°C
Assay Percent Range >98% w/v
Packaging Solid
Linear Formula C5H10O5
Quantity 25 g
Formula Weight 150.13
Chemical Name or Material D (+) Xylose
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.