Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Xylose, C.P. grade, 99% max., MP Biomedicals™

Catalog No. ICN10330090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10330090 500 g
ICN10330080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10330090 Supplier MP Biomedicals, Inc Supplier No. 0210330090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
MDL Number MFCD00064360
Synonym Wood sugar

Specifications

Color White
Assay Percent Range >99%
Quantity 500 g
Molecular Weight (g/mol) 150.13
Formula Weight 150.13
Percent Purity ≥98.5%
Grade C.P.
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Xylose
EINECSNumber 200-400-7
Recommended Storage Store at Room Temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.