Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Xylose, MP Biomedicals™

Catalog No. MP210330091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210330091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210330091 Supplier MP Biomedicals, Inc Supplier No. 0210330091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Classified as a monosaccharide of the aldopentose type. D-(+)-Xylose, MP Biomedicals is a sugar first isolated from wood. It contains five carbon atoms and includes a formyl functional group. Xylose is the main building block for hemicellulose.

  • Xylose is the main building block for hemicellulose
  • The acid-catalysed degradation of hemicellulose gives furfural, a specialty solvent in industry and a precursor to synthetic polymers
  • In animal medicine, used to test for malabsorption by administration in water after fasting
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
Synonym Wood sugar

Specifications

Melting Point 155°C to 157°C (literature)
Quantity 1 kg
Solubility Information Soluble in water (clear,colorless solution), pyridin, and hot alcohol.
Formula Weight 150.13
Percent Purity ≥98.5%
Grade Not Available
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.