Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Xylose (White to Off-White Powder), Fisher BioReagents

Catalog No. BP708250 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP708-250 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP708-250

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
D-(+)-Xylose is often used as a cell culture additive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5

Specifications

Alpha Vector Xylose
Melting Point 148°C
Color White
Assay Percent Range ≥98 %
Packaging Glass Bottle
Quantity 250 g
Merck Index 15, 10285
Specific Rotation 19 to 21° (+ or -)
Formula Weight 150.13
Percent Purity ≥98%
Physical Form Powder or Solid
Chemical Name or Material D-(+)-Xylose
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.