Learn More
Medchemexpress LLC D-Xylulose-1-13C | 131771-46-5 | 99.2% | 151.12 | 25 MG
SDP
Catalog No. 5000366737
Encompass_Preferred
D-Xylulose-1-13C is the 13C labeled D-Xylulose, a precursor of the pentiol D-arabi. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66737 Supplier Medchemexpress LLC Supplier No. 25MGHYW010256S
D-Xylulose-1-13C is the 13C labeled D-Xylulose, a precursor of the pentiol D-arabi. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.