Learn More
Medchemexpress LLC D-Xylulose-1-13C | 131771-46-5 | 99.2% | 151.12 | 5 MG
SDP
Catalog No. 5000451848
Encompass_Preferred
D-Xylulose-1-13C is a 13C labeled D-Xylulose, which serves as a precursor of the pentiol D-arabi. This compound is suitable for use as a tracer and an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes of elements like carbon are incorporated into drug molecules for quantitation during drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for laboratory chemical applications
  • Can be used in the manufacture of substances
Catalog No. 50-004-51848 Supplier Medchemexpress LLC Supplier No. HYW010256S5MG
D-Xylulose-1-13C is a 13C labeled D-Xylulose, which serves as a precursor of the pentiol D-arabi. This compound is suitable for use as a tracer and an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes of elements like carbon are incorporated into drug molecules for quantitation during drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for laboratory chemical applications
  • Can be used in the manufacture of substances