Learn More
Medchemexpress LLC D-Xylulose-13C5 | 551-84-8 | 99.9% | 155.09 | 10 MG
Supplier: Medchemexpress LLC HYW010256S310MG
D-Xylulose-13C5 is a 13C labeled D-Xylulose, which is a precursor of arabinitol. This compound serves as a valuable tool for research.
- Used as a tracer
- Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS