Learn More
Medchemexpress LLC D-Xylulose-2-13C | 131771-47-6 | 99.5% | 151.12 | 5 MG
SDP
Catalog No. 5000451855
Encompass_Preferred
D-Xylulose-2-13C is the 13C labeled form of D-Xylulose, a precursor of the pentiol D-arabi. It is useful as a tracer and an internal standard for various analytical methods.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules.
  • Can affect the pharmacokinetic and metabolic profiles of drugs through deuteration.
Catalog No. 50-004-51855 Supplier Medchemexpress LLC Supplier No. HYW010256S15MG
D-Xylulose-2-13C is the 13C labeled form of D-Xylulose, a precursor of the pentiol D-arabi. It is useful as a tracer and an internal standard for various analytical methods.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules.
  • Can affect the pharmacokinetic and metabolic profiles of drugs through deuteration.